4'-Chlorobiphenyl-4-ylboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD33401-100MGEA 7 EUR
BLDPBD33401-100MG BLDPBD33401-250MG BLDPBD33401-5G BLDPBD33401-1G
4'-Chlorobiphenyl-4-ylboronic acid 98%
4'-Chlorobiphenyl-4-ylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₀BClO₂
MW: 232,4700 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD08544389
Núm. CAS: 364044-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR