2-(4-Methylphenoxy)benzylamine hydrochloride 95%

BLDPBD333100-1GEA 106 EUR
BLDPBD333100-1G BLDPBD333100-250MG BLDPBD333100-5G BLDPBD333100-100MG
2-(4-Methylphenoxy)benzylamine hydrochloride 95%
2-(4-Methylphenoxy)benzylamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₄H₁₅NO·ClH
MW: 249,736 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03840021
Núm. CAS: 1171318-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR