1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid 95%

BLDPBD332241-100MGEA 72 EUR
BLDPBD332241-100MG BLDPBD332241-250MG
1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid 95%
1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₄N₂O₃
MW: 246,262 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09870059
Núm. CAS: 1000576-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR