5-Bromo-2-(dimethylamino)pyridine 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD32930-1GEA 10 EUR
BLDPBD32930-1G BLDPBD32930-25G BLDPBD32930-5G BLDPBD32930-10G BLDPBD32930-250MG
5-Bromo-2-(dimethylamino)pyridine 97%
5-Bromo-2-(dimethylamino)pyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₉BrN₂
MW: 201,0600 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00099486
Núm. CAS: 26163-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR