2-Amino-3-methylphenylacetic acid 95+%

BLDPBD328986-1GEA 237 EUR
BLDPBD328986-1G BLDPBD328986-100MG BLDPBD328986-250MG
2-Amino-3-methylphenylacetic acid 95+%
2-Amino-3-methylphenylacetic acid
Formula: C₉H₁₁NO₂
MW: 165,189 g/mol
Número MDL: MFCD16606085
Núm. CAS: 1334405-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR