4-Fluoro-4'-nitrobenzophenone 98%

BLDPBD3231-25GEA 306 EUR
BLDPBD3231-25G BLDPBD3231-5G BLDPBD3231-1G
4-Fluoro-4'-nitrobenzophenone 98%
4-Fluoro-4'-nitrobenzophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₈FNO₃
MW: 245,2059 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00053428
Núm. CAS: 2195-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR