2-Methoxy-5-(trifluoromethoxy)aniline 98%

BLDPBD320557-100MGEA 25 EUR
BLDPBD320557-100MG BLDPBD320557-5G BLDPBD320557-250MG BLDPBD320557-1G BLDPBD320557-10G
2-Methoxy-5-(trifluoromethoxy)aniline 98%
2-Methoxy-5-(trifluoromethoxy)aniline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₈F₃NO₂
MW: 207,15 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09832305
Núm. CAS: 660848-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR