1-Bromo-4-(isopropylthio)benzene 96%

Proveedor: BLD PHARMATECH GMBH

BLDPBD315820-100MGEA 15 EUR
BLDPBD315820-100MG BLDPBD315820-1G BLDPBD315820-250MG
1-Bromo-4-(isopropylthio)benzene 96%
1-Bromo-4-(isopropylthio)benzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₁BrS
MW: 231,153 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD20484423
Núm. CAS: 70398-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR