4-(3-Fluorophenoxy)piperidine hydrochloride 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD315545-1GEA 269 EUR
BLDPBD315545-1G BLDPBD315545-5G BLDPBD315545-250MG BLDPBD315545-100MG
4-(3-Fluorophenoxy)piperidine hydrochloride 98%
4-(3-Fluorophenoxy)piperidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₅ClFNO
MW: 231,6900 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08445760
Núm. CAS: 3202-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR