2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

BLDPBD314532-5GEA 154 EUR
BLDPBD314532-5G BLDPBD314532-1G BLDPBD314532-250MG
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₂₁BO₂
MW: 232,126 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD18383605
Núm. CAS: 1232132-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR