2-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-fluoren-9-one 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD314070-1GEA 45 EUR
BLDPBD314070-1G BLDPBD314070-250MG BLDPBD314070-5G
2-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-fluoren-9-one 95%
2-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-fluoren-9-one

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₉H₁₉BO₃
MW: 306,163 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD16294551
Núm. CAS: 1021306-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR