Potassium-4-(hydroxymethyl)phenyltrifluoroborate 98+%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD314033-1GEA
17
EUR
BLDPBD314033-1G
BLDPBD314033-0.025
BLDPBD314033-5G
BLDPBD314033-25G
Potassium-4-(hydroxymethyl)phenyltrifluoroborate 98+%
Potassium-4-(hydroxymethyl)phenyltrifluoroborate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₇H₇BF₃KO MW: 214,04 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08276800 Núm. CAS: 1015082-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...