1,3-Dibromo-2,5-difluorobenzene 97%

BLDPBD307232-25GEA 387 EUR
BLDPBD307232-25G BLDPBD307232-1G BLDPBD307232-5G BLDPBD307232-250MG
1,3-Dibromo-2,5-difluorobenzene 97%
1,3-Dibromo-2,5-difluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₂Br₂F₂
MW: 271,8900 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09878120
Núm. CAS: 128259-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR