4-Bromo-5-chloro-2-fluoroaniline 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD307118-1GEA 33 EUR
BLDPBD307118-1G BLDPBD307118-250MG BLDPBD307118-5G
4-Bromo-5-chloro-2-fluoroaniline 98%
4-Bromo-5-chloro-2-fluoroaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₄BrClFN
MW: 224,4600 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09878173
Núm. CAS: 116369-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR