Nile red 98%

BLDPBD306389-1GEA 146 EUR
BLDPBD306389-1G BLDPBD306389-5G BLDPBD306389-250MG
Nile red 98%
Nile red

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₀H₁₈N₂O₂
MW: 318,38 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011639
Núm. CAS: 7385-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR