3-Chloro-1H-indazole-5-carboxylic acid 95+%

Proveedor: BLD PHARMATECH GMBH

BLDPBD303212-250MGEA 168 EUR
BLDPBD303212-250MG BLDPBD303212-1G BLDPBD303212-100MG
3-Chloro-1H-indazole-5-carboxylic acid 95+%
3-Chloro-1H-indazole-5-carboxylic acid
Formula: C₈H₅ClN₂O₂
MW: 196,591 g/mol
Número MDL: MFCD11499013
Núm. CAS: 1031417-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR