2,5-Difluoro-p-toluic acid 97%

BLDPBD29991-5GEA 59 EUR
BLDPBD29991-5G BLDPBD29991-250MG
2,5-Difluoro-p-toluic acid 97%
2,5-Difluoro-p-toluic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₈H₆F₂O₂
MW: 172,13 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD08063230
Núm. CAS: 103877-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR