1H-Indazole-6-boronic acid 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD29812-25GEA 569 EUR
BLDPBD29812-25G BLDPBD29812-10G BLDPBD29812-1G BLDPBD29812-5G BLDPBD29812-250MG
1H-Indazole-6-boronic acid 97%
1H-Indazole-6-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₇BN₂O₂
MW: 161,9500 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD04972419
Núm. CAS: 885068-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR