5-Chloro-2-formylphenylboronic acid 95%

BLDPBD29783-5GEA 101 EUR
BLDPBD29783-5G BLDPBD29783-100MG BLDPBD29783-25G BLDPBD29783-250MG BLDPBD29783-1G
5-Chloro-2-formylphenylboronic acid 95%
5-Chloro-2-formylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₆BClO₃
MW: 184,3900 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03425955
Núm. CAS: 870238-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR