3-(4-Chlorophenyl)-1H-pyrazol-5-amine 98%

BLDPBD29627-1GEA 102 EUR
BLDPBD29627-1G BLDPBD29627-5G BLDPBD29627-250MG BLDPBD29627-100MG
3-(4-Chlorophenyl)-1H-pyrazol-5-amine 98%
3-(4-Chlorophenyl)-1H-pyrazol-5-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₈ClN₃
MW: 193,64 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00053046
Núm. CAS: 78583-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR