5-Amino-1H-pyrazolo[4,3-b]pyridine 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD295188-250MGEA 138 EUR
BLDPBD295188-250MG BLDPBD295188-1G BLDPBD295188-100MG
5-Amino-1H-pyrazolo[4,3-b]pyridine 98%
5-Amino-1H-pyrazolo[4,3-b]pyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₆N₄
MW: 134,139 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD22550615
Núm. CAS: 1206974-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR