Ethyl-2,3,4-trifluorobenzoate 97%

BLDPBD292247-1GEA 6 EUR
BLDPBD292247-1G BLDPBD292247-25G BLDPBD292247-5G
Ethyl-2,3,4-trifluorobenzoate 97%
Ethyl-2,3,4-trifluorobenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₇F₃O₂
MW: 204,1459 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06204408
Núm. CAS: 351354-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR