Tetrakis(dimethylsiloxy)silane 96%

BLDPBD290640-25GEA 71 EUR
BLDPBD290640-25G BLDPBD290640-5G BLDPBD290640-100G
Tetrakis(dimethylsiloxy)silane 96%
Tetrakis(dimethylsiloxy)silane

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₂₈O₄Si₅
MW: 328,73 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00053848
Núm. CAS: 17082-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR