Methyl-3-aminoisonicotinate 98%
|
|
BLDPBD29036-500GEA
1529
EUR
BLDPBD29036-500G
BLDPBD29036-100G
BLDPBD29036-25G
BLDPBD29036-1G
BLDPBD29036-10G
BLDPBD29036-5G
Methyl-3-aminoisonicotinate 98%
Methyl-3-aminoisonicotinate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₇H₈N₂O₂ MW: 152,20 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02082535 Núm. CAS: 55279-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...