Methyl-3-fluoro-4-iodobenzoate 98%

BLDPBD287712-250MGEA 23 EUR
BLDPBD287712-250MG BLDPBD287712-1G BLDPBD287712-5G
Methyl-3-fluoro-4-iodobenzoate 98%
Methyl-3-fluoro-4-iodobenzoate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₆FIO₂
MW: 280,0349 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09800728
Núm. CAS: 1027513-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR