Methyl-1H-indazole-3-carboxylate 98%
|
|
BLDPBD28720-500GEA
606
EUR
BLDPBD28720-500G
BLDPBD28720-100G
BLDPBD28720-10G
BLDPBD28720-5G
BLDPBD28720-25G
BLDPBD28720-1G
Methyl-1H-indazole-3-carboxylate 98%
Methyl-1H-indazole-3-carboxylate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₈N₂O₂ MW: 176,1700 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01138133 Núm. CAS: 43120-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...