(4-Benzylmorpholin-2-yl)methanol 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD28669-1GEA 37 EUR
BLDPBD28669-1G BLDPBD28669-10G BLDPBD28669-250MG BLDPBD28669-5G
(4-Benzylmorpholin-2-yl)methanol 98%
(4-Benzylmorpholin-2-yl)methanol

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₇NO₂
MW: 207,269 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02682009
Núm. CAS: 40987-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR