5-Amino-2-bromobenzoic acid 98%
|
|
BLDPBD28258-100GEA
224
EUR
BLDPBD28258-100G
BLDPBD28258-10G
BLDPBD28258-1G
BLDPBD28258-25G
BLDPBD28258-5G
5-Amino-2-bromobenzoic acid 98%
5-Amino-2-bromobenzoic acid
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₇H₆BrNO₂ MW: 216,03 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07368968 Núm. CAS: 2840-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...