2-Amino-4-chloro-5-nitropyridine 96%
|
|
BLDPBD28150-250MGEA
11
EUR
BLDPBD28150-0.025G
BLDPBD28150-250MG
BLDPBD28150-10G
BLDPBD28150-1G
BLDPBD28150-25G
BLDPBD28150-5G
2-Amino-4-chloro-5-nitropyridine 96%
2-Amino-4-chloro-5-nitropyridine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₅H₄ClN₃O₂ MW: 173,557 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD04973545 Núm. CAS: 24484-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...