2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%

Proveedor: BLD PHARMATECH GMBH

BLDPBD27775-10GEA 293 EUR
BLDPBD27775-10G BLDPBD27775-5G BLDPBD27775-25G BLDPBD27775-250MG BLDPBD27775-1G
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₃H₁₉N₃O₂
MW: 369,42 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01863586
Núm. CAS: 174500-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR