Quinoxaline-5-carboxaldehyde 95%

BLDPBD27064-1GEA 181 EUR
BLDPBD27064-1G BLDPBD27064-100MG BLDPBD27064-5G BLDPBD27064-250MG
Quinoxaline-5-carboxaldehyde 95%
Quinoxaline-5-carboxaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₆N₂O
MW: 158,157 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00805833
Núm. CAS: 141234-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR