1H-Indazole-5-boronic acid pinacol ester 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD26874-250MGEA 29 EUR
BLDPBD26874-250MG BLDPBD26874-5G BLDPBD26874-1G BLDPBD26874-25G BLDPBD26874-10G
1H-Indazole-5-boronic acid pinacol ester 97%
1H-Indazole-5-boronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₃H₁₇BN₂O₂
MW: 244,0973 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD07367523
Núm. CAS: 862723-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR