Ethyl-1H-indazole-6-carboxylate 98%
|
|
BLDPBD26786-1GEA
62
EUR
BLDPBD26786-1G
BLDPBD26786-250MG
Ethyl-1H-indazole-6-carboxylate 98%
Ethyl-1H-indazole-6-carboxylate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₀H₁₀N₂O₂ MW: 190,2000 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07371613 Núm. CAS: 713-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...