5-Methoxypyridine-3-boronic acid 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD26319-1GEA 47 EUR
BLDPBD26319-1G BLDPBD26319-25G BLDPBD26319-5G
5-Methoxypyridine-3-boronic acid 95%
5-Methoxypyridine-3-boronic acid
Formula: C₆H₈BNO₃
MW: 152,95 g/mol
Número MDL: MFCD03425977
Núm. CAS: 850991-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR