4-(2-Carboxyethyl)phenylboronic acid pinacol ester 98%
|
|
BLDPBD260917-1GEA
54
EUR
BLDPBD260917-1G
BLDPBD260917-250MG
BLDPBD260917-5G
4-(2-Carboxyethyl)phenylboronic acid pinacol ester 98%
4-(2-Carboxyethyl)phenylboronic acid pinacol ester
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₅H₂₁BO₄ MW: 276,1400 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11053827 Núm. CAS: 797755-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...