2-Amino-4-(2-pyridyl)thiazole 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD26041-5GEA 245 EUR
BLDPBD26041-5G BLDPBD26041-10G BLDPBD26041-250MG BLDPBD26041-1G
2-Amino-4-(2-pyridyl)thiazole 97%
2-Amino-4-(2-pyridyl)thiazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₇N₃S
MW: 177,22 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00460417
Núm. CAS: 30235-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR