Octahydro-4H-isoindol-4-one, N-BOC protected 98%

BLDPBD258911-100MGEA 84 EUR
BLDPBD258911-100MG BLDPBD258911-250MG BLDPBD258911-1G
Octahydro-4H-isoindol-4-one, N-BOC protected 98%
Octahydro-4H-isoindol-4-one, N-BOC protected

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₂₁NO₃
MW: 239,311 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD17170012
Núm. CAS: 879687-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR