4-Chloro-N,N-dimethylaniline 97%

BLDPBD258147-250MGEA 19 EUR
BLDPBD258147-250MG BLDPBD258147-1G BLDPBD258147-10G BLDPBD258147-5G BLDPBD258147-25G
4-Chloro-N,N-dimethylaniline 97%
4-Chloro-N,N-dimethylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₁₀ClN
MW: 155,625 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07780200
Núm. CAS: 698-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR