5-Nitro-2-thiophenecarboxylic acid 95%

BLDPBD2570-100GEA 173 EUR
BLDPBD2570-100G BLDPBD2570-5G BLDPBD2570-500G BLDPBD2570-25G BLDPBD2570-1G BLDPBD2570-10G
5-Nitro-2-thiophenecarboxylic acid 95%
5-Nitro-2-thiophenecarboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₃NO₄S
MW: 173,14 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00159552
Núm. CAS: 6317-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR