1-Chloro-4-iodo-2,7-naphthyridine 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD256958-250MGEA 356 EUR
BLDPBD256958-250MG BLDPBD256958-100MG BLDPBD256958-1G
1-Chloro-4-iodo-2,7-naphthyridine 95%
1-Chloro-4-iodo-2,7-naphthyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₈H₄ClIN₂
MW: 290,488 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD17015945
Núm. CAS: 1234616-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR