Ethyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate 97%

BLDPBD25288-1GEA 24 EUR
BLDPBD25288-1G BLDPBD25288-250MG BLDPBD25288-5G
Ethyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₂N₂O₂
MW: 204,2300 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00139509
Núm. CAS: 2549-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR