2-Iodo-5-trifluoromethylpyridine 98%

BLDPBD25245-100GEA 542 EUR
BLDPBD25245-100G BLDPBD25245-5G BLDPBD25245-25G BLDPBD25245-250MG BLDPBD25245-1G
2-Iodo-5-trifluoromethylpyridine 98%
2-Iodo-5-trifluoromethylpyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₃F₃IN
MW: 272,99 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08741351
Núm. CAS: 100366-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR