4-Fluoro-o-anisidine hydrochloride 98%
|
|
BLDPBD251493-5GEA
133
EUR
BLDPBD251493-5G
BLDPBD251493-25G
BLDPBD251493-250MG
BLDPBD251493-1G
4-Fluoro-o-anisidine hydrochloride 98%
4-Fluoro-o-anisidine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₇H₉ClFNO MW: 177,6000 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09800850 Núm. CAS: 178671-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...