Diethyl (1-ethoxyethylidene)malonate 95%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD244336-100MGEA
23
EUR
BLDPBD244336-100MG
BLDPBD244336-5G
BLDPBD244336-1G
BLDPBD244336-250MG
Diethyl (1-ethoxyethylidene)malonate 95%
Diethyl (1-ethoxyethylidene)malonate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₁H₁₈O₅ MW: 230,258 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD22575006 Núm. CAS: 3044-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...