4-Hydroxy-1-methyl-1H-pyrazole 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD241017-5GEA 546 EUR
BLDPBD241017-5G BLDPBD241017-1G BLDPBD241017-250MG BLDPBD241017-100MG
4-Hydroxy-1-methyl-1H-pyrazole 95%
4-Hydroxy-1-methyl-1H-pyrazole

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₄H₆N₂O
MW: 98,103 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD22414500
Núm. CAS: 78242-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR