4-(Trifluoromethyl)-N,N-dimethylbenzamide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD240094-1GEA 42 EUR
BLDPBD240094-1G BLDPBD240094-25G BLDPBD240094-5G BLDPBD240094-10G BLDPBD240094-250MG
4-(Trifluoromethyl)-N,N-dimethylbenzamide 98%
4-(Trifluoromethyl)-N,N-dimethylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀F₃NO
MW: 217,1877 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD16618942
Núm. CAS: 25771-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR