Methyl 5-bromo-2-methoxyisonicotinate 97%

BLDPBD239032-100MGEA 61 EUR
BLDPBD239032-100MG
Methyl 5-bromo-2-methoxyisonicotinate 97%
Methyl 5-bromo-2-methoxyisonicotinate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈BrNO₃
MW: 246,058 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07375112
Núm. CAS: 886365-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR