4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD237841-100GEA 533 EUR
BLDPBD237841-100G BLDPBD237841-10G BLDPBD237841-250MG BLDPBD237841-1G BLDPBD237841-5G BLDPBD237841-25G
4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%
4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₇H₂₅BO₄
MW: 304,1890 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03453055
Núm. CAS: 850568-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR