3'-Bromo-5'-fluoroacetophenone 97%
|
|
BLDPBD237441-1GEA
30
EUR
BLDPBD237441-1G
BLDPBD237441-25G
BLDPBD237441-100MG
BLDPBD237441-250MG
BLDPBD237441-5G
3'-Bromo-5'-fluoroacetophenone 97%
3'-Bromo-5'-fluoroacetophenone
Storage condition: Sealed in dry, 2-8°C
Formula:
C₈H₆BrFO MW: 217,04 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11847746 Núm. CAS: 105515-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...