2,3,4-Trifluoroiodobenzene 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD236695-1GEA 9 EUR
BLDPBD236695-1G BLDPBD236695-10G BLDPBD236695-5G BLDPBD236695-100G BLDPBD236695-25G
2,3,4-Trifluoroiodobenzene 97%
2,3,4-Trifluoroiodobenzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₂F₃I
MW: 257,9798 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03788551
Núm. CAS: 459424-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR